Learn More
TG101348, TRC
Specifications
Specifications
| Chemical Name or Material | TG101348 |
| CAS | 936091-26-8 |
| Molecular Formula | C27 H36 N6 O3 S |
| Quantity | 5 mg |
| Synonym | Benzenesulfonamide','N-(1,1-dimethylethyl)-3-[[5-methyl-2-[[4-[2-(1-pyrrolidinyl)ethoxy]phenyl]amino]-4-pyrimidinyl]amino]-, N-(1,1-Dimethylethyl)-3-[[5-methyl-2-[[4-[2-(1-pyrrolidinyl)ethoxy]phenyl]amino]-4-pyrimidinyl]amino]benzenesulfonamide, Fedratinib, Inrebic, SAR 302503, TG 101348, TG 10138, N-tert-Butyl-3-[[5-methyl-2-[4-[2-(pyrrolidin-1-yl)ethoxy]anilino]pyrimidin-4-yl]amino]benzenesulfonamide |
| InChI Formula | InChI=1S/C27H36N6O3S/c1-20-19-28-26(30-21-10-12-23(13-11-21)36-17-16-33-14-5-6-15-33)31-25(20)29-22-8-7-9-24(18-22)37(34,35)32-27(2,3)4/h7-13,18-19,32H,5-6,14-17H2,1-4H3,(H2,28,29,30,31) |
| SMILES | Cc1cnc(Nc2ccc(OCCN3CCCC3)cc2)nc1Nc4cccc(c4)S(=O)(=O)NC(C)(C)C |
| IUPAC Name | N-tert-butyl-3-[[5-methyl-2-[4-(2-pyrrolidin-1-ylethoxy)anilino]pyrimidin-4-yl]amino]benzenesulfonamide |
| Molecular Weight (g/mol) | 524.68 |
| Formula Weight | 524.257 |