missing translation for 'onlineSavingsMsg'
Learn More

TG101209, MedChemExpress

Product Code. p-200097196
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453958

Brand: Medchem Express HY10410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TG101209 is a selective JAK2 inhibitor with IC50 of 6 nM, less potent to Flt3 and RET with IC50 of 25 nM and 17 nM, appr 30-fold selective for JAK2 than JAK3, and sensitive to JAK2V617F and MPLW515L/K mutations.

TG101209 is a selective JAK2 inhibitor with IC50 of 6 nM, less potent to Flt3 and RET with IC50 of 25 nM and 17 nM, appr 30-fold selective for JAK2 than JAK3, and sensitive to JAK2V617F and MPLW515L/K mutations.

Specifications

Chemical Name or Material TG101209
CAS 936091-14-4
Color Gray
Purity Grade Notes Research
Molecular Formula C26H35N7O2S
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (98.10 mM)
SMILES O=S(C1=CC=CC(NC2=NC(NC3=CC=C(C=C3)N4CCN(CC4)C)=NC=C2C)=C1)(NC(C)(C)C)=O
Molecular Weight (g/mol) 509.67
Formula Weight 509.67
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.