Learn More
TG 100572 Hydrochloride, MedChemExpress
Product Code. 16483608
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483608 10 mg 10mg
16440929 1 mg 1mg
16420007 50 mg 50mg
16456337 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16483608 Supplier Medchem Express Supplier No. HY10185
 Restricted Item
Estimated Shipment: 27-11-2026
MedChemExpress TG 100572 Hydrochloride is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases; has IC50s of 2, 7, 2, 16, 13, 5, 0.5, 6, 0.1, 0.4, 1, 0.2 nM for VEGFR1, VEGFR2, FGFR1, FGFR2, PDGFRβ, Fgr, Fyn, Hck, Lck, Lyn, Src, Yes, respectively.
TG 100572 Hydrochloride is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases
Specifications
Chemical Name or Material TG 100572 Hydrochloride
CAS 867331-64-4
Color Red
Purity Grade Notes Research
Molecular Formula C26H27Cl2N5O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (48.79 mM; Need ultrasonic)
SMILES ClC1=CC=C(O)C=C1C2=CC(C)=C(N=C(NC(C=C3)=CC=C3OCCN4CCCC4)N=N5)C5=C2.Cl
Molecular Weight (g/mol) 512.43
Formula Weight 512.43
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation