missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TFF1 Recombinant Protein

Product Code. 16111342
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16111342 10 μg 10µg
16121342 25 μg 25µg
2 options
This item is not returnable. View return policy

Product Code. 16111342

Brand: Abnova™ H00007031P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human TFF1 full-length ORF recombinant protein with GST-tag at N-terminal

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.

  • Molecular Weight: 34.98kDa
  • Preparation Method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer

Sequence: MATMENKVICALVLVSMLALGTLAEAQTETCTVAPRERQNCGFPGVTPSQCANKGCCFDDTVRGVPWCFYPNTIDVPPEEECEF

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH32811.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 7031
Molecular Weight (g/mol) 34.98
Name TFF1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MATMENKVICALVLVSMLALGTLAEAQTETCTVAPRERQNCGFPGVTPSQCANKGCCFDDTVRGVPWCFYPNTIDVPPEEECEF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias BCEI/D21S21/HP1.A/HPS2/pNR-2/pS2
Common Name TFF1
Gene Symbol TFF1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.