Learn More
TFAP2A AP-2 Mouse anti-Human, Unconjugated, Clone: 1B12F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67076-1-IG-20UL
Description
The TFAP2A, AP-2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFAP2A, AP-2. This antibody recognizes human, mouse, rat antigen. The TFAP2A, AP-2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| TFAP2A AP-2 | |
| Monoclonal | |
| 900 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 1B12F11 | |
| Unconjugated | |
| 20 μL | |
| 7020 | |
| IgG1 |