Learn More
TFAP2A AP-2 Mouse anti-Human, Unconjugated, Clone: 1B12F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67076-1-IG-20UL
 View more versions of this product
Product Code. 16809495
  • 1710.33 NOK / 20µL
Description
The TFAP2A, AP-2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFAP2A, AP-2. This antibody recognizes human, mouse, rat antigen. The TFAP2A, AP-2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TFAP2A AP-2
Monoclonal
900 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
1B12F11
Unconjugated
20 μL
7020
IgG1