Learn More
Texwipe™ Isolator Cleaning Tool Replacement Telescoping Handle
Product Code. 17955241 Shop All Texwipe Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Autoclavable Unit Size
17955241 Autoclavable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17955241 Supplier Texwipe™ Supplier No. TX7122
  • ONLINE EXCLUSIVE PRICE
    935.00 NOK
    761.98 NOK / Pack of 1
    Save 173.02 NOK
    18% Off
Estimated Shipment: 04-03-2027
Specifications
Product Type Replacement Telescoping Handle
Color White
Autoclavable Autoclavable
For Use With (Equipment) Isolator Cleaning Tool
Material Fiberglass