Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
Product Code. 11575315 Shop All Texwipe Products
Change view
Click to view available options
Color:
White
Dimensions (L x W):
45 x 25 in.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Unit Size
11575315 White 45 x 25 in. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11575315 Supplier Texwipe™ Supplier No. AMA254582W
  • ONLINE EXCLUSIVE PRICE
    3655.00 NOK
    3305.77 NOK / Pack of 4
    Save 349.23 NOK
    9% Off
Estimated Shipment: 09-12-2026
Apply directly to floor
Choose 30- or 60-layer mats.
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Dimensions (L x W) 45 x 25 in.
Quantity 60 Sheets/Mat
Color White