Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Product Code. 11565315 Shop All Texwipe Products
Change view
Click to view available options
Color:
Blue
Gray
White
Dimensions (L x W):
25 x 45 in.
45 x 25 in.
46 x 36 in.
Unit Size:
Pack of 8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) unitSize
11565315 White 45 x 25 in. Pack of 8
11525325 Blue 46 x 36 in. Pack of 8
11505325 Blue 45 x 25 in. Pack of 8
12410070 White 46 x 36 in. Pack of 8
15762386 Gray 25 x 45 in. Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11565315 Supplier Texwipe™ Supplier No. AMA254581W
  • ONLINE EXCLUSIVE PRICE
    3935.00 NOK
    3562.08 NOK / Pack of 8
    Save 372.92 NOK
    9% Off
View all promotions
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Apply directly to floor
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
Specifications
Dimensions (L x W) 45 x 25 in.
Quantity 30 Sheets/Mat
Color White