Learn More
Texwipe™ AlphaMop™ Replacement Microdenier Mop Covers
Shop All Texwipe Products
Click to view available options
:
For use with AlphaMop and Autoclavable AlphaMop; Dim.: 8 x 15 in.
Specifications
Specifications
| Product Type | Mop Cover |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Includes | Foam Pads |
| For Use With (Application) | Maximum removal of small and micro contaminants and used in cleanrooms, disinfecting with quat-based cleaners, pharmaceuticals, biologics, medical device |
| Material | Microdenier Polyester Fiber |