Learn More
Texwipe™ AlphaMop™ Replacement Handle
Shop All Texwipe Products
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Each
Description
Specifications
Specifications
| Product Type | Replacement Handle |
| Color | White |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| For Use With (Equipment) | AlphaMop |
| Color-coded | No |
| Material | Fiberglass |