Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
Shop All Texwipe Products
Click to view available options
Product Type:
Mop
Unit Size:
Each
Description
Includes
Two handles (18 and 24 in. long), one polyester foam pad, and six TechniCloth™ polyester mop covers. Mop with telescoping handle includes six polyester mop covers.
- Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
- Made from autoclavable Celcon™ plastic
- Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Specifications
| Product Type | Mop |
| Autoclavable | Autoclavable |
| Material | Plastic |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu