Description
- These single-ply wipers have the liquid-holding capacity of two-ply wipers
- TexTra™ combines this high absorbency with great abrasion resistance
- Can be autoclaved for sterile or aseptic environments
Specifications
Specifications
| Certifications/Compliance | ISO Class 3 – 8, Class 1 – 100,000, EU Grade A – D |
| Dimensions (L x W) | 25 x 60 cm |
| Sterility | Sterile |