Learn More
Tetramethyltin, Sigma-Aldrich™
Specifications
Specifications
| CAS | 594-27-4 |
| Melting Point | -54°C |
| Density | 1.29 g/cm3 at 20°C |
| Boiling Point | 74°C to 75°C/1013 hPa |
| Molecular Formula | Sn(CH3)4 |
| MDL Number | MFCD00008278 |
| Quantity | 10 mL |
| Synonym | Tetramethyltin, Tetramethylstannane, Tin tetramethyl, Tetramethyltin |
| Molecular Weight (g/mol) | 178.85 |
| Grade | Research |