Learn More
Tetramethylammonium hydroxide, 25% in water
Product Code. p-198388 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
5kg
Product Code. 10465281
Brand: Thermo Scientific Acros 420521000
Request Bulk or Custom Format
CAS: 75-59-2 | C4H13NO | 91.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetramethylammonium hydroxide
Name Note 25% in Water
CAS 75-59-2, 7732-18-5
Melting Point -25.0°C
Color Colorless to Yellow
Density 1.0140g/mL
Boiling Point 102.0°C
Flash Point >95°C
Packaging Plastic bottle
Molecular Formula C4H13NO
Linear Formula (CH3)4NOH
Refractive Index 1.3780 to 1.3840 (20°C, 589nm)
MDL Number MFCD00008280
Quantity 100 g
Beilstein 04,5
Fieser 11,514
Merck Index 15,9371
Specific Gravity 1.014
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
Solubility Information Solubility in water: soluble.
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.15
PubChem CID 60966
Viscosity 3.13 mPa.s (19°C)
Formula Weight 91.15
Percent Purity 23 to 27%
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium hydroxide, 25% in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetramethylammoniumhydroxide