missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl Succinonitrile, MP Biomedicals

Product Code. 11458705 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11458705 Supplier MP Biomedicals Supplier No. 0522460501

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3333-52-6
Molecular Formula C8H12N2
MDL Number MFCD00040683
Quantity 1 g
Synonym tetramethylsuccinonitrile, butanedinitrile, tetramethyl, 2,2,3,3-tetramethylsuccinonitrile, tetramethylsuccinotrile, tetramethyl succinonitrile, tetramethylsuccinodinitrile, tmsn, succinonitrile, tetramethyl, tetramethylbutanedinitrile, tetramethylsukcinonitril
InChI Key ZVQXQPNJHRNGID-UHFFFAOYSA-N
SMILES CC(C)(C#N)C(C)(C)C#N
IUPAC Name tetramethylbutanedinitrile
Molecular Weight (g/mol) 136.20
PubChem CID 18745
Formula Weight 136.2
Chemical Name or Material Tetramethyl Succinonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.