Learn More
Tetrakis(triphenylphosphine)-palladium(0), Sigma-Aldrich™
Description
Tetrakis(triphenylphosphine)-palladium(0) can be used to catalyze the formation of carbon-carbon bonds, the reduction of functional groups, the formation of carbon-metal bonds, and the deprotection of allyloxycarbonyl groups.
Specifications
Specifications
| CAS | 14221-01-3 |
| Melting Point | 103°C to 107°C |
| Molecular Formula | Pd(P(C6H5)3)4 |
| MDL Number | MFCD00010012 |
| Quantity | 5 g |
| Synonym | Tetrakis(triphenylphosphine)-palladium(0), Tetrakis(triphenylphosphine)-palladium(0) |
| Molecular Weight (g/mol) | 1155.56 |
| Grade | Research |