missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Tetrahydrofurfuryl alcohol, 98%

Product Code. 11478946
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11478946

Brand: Thermo Scientific Alfa Aesar A17078.0F

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-99-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00005372
InChI Key BSYVTEYKTMYBMK-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
PubChem CID 7360
SMILES OCC1CCCO1

Specifications

Melting Point <-80°C
Density 1.053
Boiling Point 178°C
Quantity 2500 mL
Refractive Index 1.452
Beilstein 102723
Merck Index 14,9213
IUPAC Name oxolan-2-ylmethanol
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material (+/-)-Tetrahydrofurfuryl alcohol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.