Learn More
Tetrahydrofuran-d8, TRC
Product Code. 30416889
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416889 500 mg
30424056 250 mg
30422832 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30416889 Supplier Toronto Research Chemicals Supplier No. TRCT293512500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1693-74-9
Molecular Formula C4 2H8 O
Molecular Weight (g/mol) 80.16
Synonym Furan-d4, tetrahydro-d4- (6CI,7CI,8CI,9CI), Tetrahydro-d4furan-2,3,4,5-d4, Perdeuterotetrahydrofuran, THF-d8, Tetrahydrofuran-d8, d8-THF
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C4H8O/c1-2-4-5-3-1/h1-4H2/i1D2,2D2,3D2,4D2
Formula Weight 80.1077
Chemical Name or Material Tetrahydrofuran D8