Learn More
Tetrahydrodicyclopentadiene, TRC
Specifications
Specifications
| Chemical Name or Material | Tetrahydrodicyclopentadiene |
| CAS | 6004-38-2 |
| Molecular Formula | C10H16 |
| Quantity | 1 g |
| Synonym | Octahydro-4,7-methano-1H-indene, 2,3-Trimethylenenorbornane, 2,3-(1,3-Propanediyl)bicyclo[2.2.1]heptane, Hexahydro-4,7-methanoindan, NSC 22464, Tetrahydrocyclopentadiene dimer, Tricyclo[5.2.1.02,6]decane, Trimethylenenorbornane |
| InChI Formula | InChI=1S/C10H16/c1-2-9-7-4-5-8(6-7)10(9)3-1/h7-10H,1-6H2 |
| SMILES | C1(CCC2)C2C3CC1CC3 |
| Molecular Weight (g/mol) | 136.23 |
| Formula Weight | 136.1252 |
Product Title
Hittar du en möjlighet till förbättring?