Learn More
Tetraethyl Orthotitanate, Sigma-Aldrich™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This versatile Lewis acid catalyzes aldol condensations, transesterifications, and Claisen rearrangements with high activity. Its ability to coordinate oxygen-containing groups enables stereoselective synthesis of complex molecules.
Specifications
Specifications
| CAS | 3087-36-3 |
| Melting Point | -40°C |
| Density | 1.08 g/cm3 at 20°C |
| Boiling Point | 150°C to 155°C/16 hPa |
| Molecular Formula | (C2H5O)4Ti |
| MDL Number | MFCD00009071 |
| Quantity | 250 mL |
| Synonym | Tetraethyl orthotitanate, Tetraethyl orthotitanate |
| Molecular Weight (g/mol) | 228.11 |
| Grade | Research |