missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyltrimethylammonium Bromide, Sigma-Aldrich™

Artikelnummer. 15871841
Ansicht ändern
Click to view available options
Quantity:
25 g
Packungsgröße:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Unit Size
15871841 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15871841 Supplier Sigma Aldrich Supplier No. 8400060025
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 1119-97-7
Melting Point 245°C to 250°C
MDL Number MFCD00011770
Quantity 25 g
Synonym Tetradecyltrimethylammonium bromide, N,N,N-Trimethyl-1-tetradecanammonium bromide, Myristyltrimethylammonium bromide, Tetradecyltrimethylammonium bromide
Molecular Weight (g/mol) 336.39
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.