Learn More
Tetradecanonitrile, 98%, Thermo Scientific™
Product Code. 11428597
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428597 100 g 100g
11418597 25 g 25g
11438597 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11428597 Supplier Thermo Scientific Alfa Aesar Supplier No. A19704.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 629-63-0
Molecular Formula C14H27N
Molecular Weight (g/mol) 209.38
MDL Number MFCD00019848
InChI Key MLRCLPRHEOPXLL-UHFFFAOYSA-N
Synonym myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3
PubChem CID 64678
IUPAC Name tetradecanenitrile
SMILES CCCCCCCCCCCCCC#N
Specifications
Melting Point ∼19°C
Density 0.83
Boiling Point 226°C to 227°C (100 mmHg)
Odor Odorless
Refractive Index 1.4415
Quantity 100 g
Beilstein 1705598
Formula Weight 209.38
Percent Purity 98%
Chemical Name or Material Tetradecanonitrile