Learn More
Tetracosanol, 99+%
Product Code. p-3758774 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
Product Code. Quantity Packaging unitSize
10095673 100 mg Glass bottle 100mg
10114820 1 g Glass bottle 1g
2 options
Product Code. 10095673
Brand: Thermo Scientific Acros 258151000
Request Bulk or Custom Format
CAS: 506-51-4 | C24H50O | 354.66 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 506-51-4
Molecular Formula C24H50O
Molecular Weight (g/mol) 354.66
MDL Number MFCD00042662
InChI Key TYWMIZZBOVGFOV-UHFFFAOYSA-N
Synonym 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc
PubChem CID 10472
ChEBI CHEBI:77413
IUPAC Name tetracosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCO
Specifications
Melting Point 75°C to 77°C
Color White
Flash Point 218°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)23OH
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ether,ketones,,specially when hot
Formula Weight 354.65
Percent Purity 99+%
Physical Form Shiny Flakes or Fluffy Powder
Chemical Name or Material Tetracosanol