missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide, 0.1 M, in 2-propanol/methanol, Scharlab

Product Code. 11382565
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382565 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11382565 Supplier Scharlab Supplier No. TE01161000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2052-49-5
Molecular Formula C16H37NO
MDL Number MFCD00009425
Quantity 1 L
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium hydroxide
Molecular Weight (g/mol) 259.48
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide, solution 0,1 mol/L, in 2-propanolol/methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H371-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol, tetrabutylammonium hydroxide, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.