Learn More
Tetrabromophthalic Anhydride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 632-79-1 |
| Melting Point | 271°C to 274°C |
| Density | 2.87 g/cm3 at 20°C |
| MDL Number | MFCD00005919 |
| Quantity | 250 g |
| Synonym | Tetrabromophthalic anhydride, Tetrabromophthalic anhydride |
| Molecular Weight (g/mol) | 463.7 |
| Grade | Research |