Learn More
Tetrabenzyl Pyrophosphate, TRC
Product Code. 30412593
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412593 1 g -
30428677 2.5 g 2.5g
30424115 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30412593 Supplier Toronto Research Chemicals Supplier No. TRCT2894501G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 990-91-0
Molecular Formula C28 H28 O7 P2
Molecular Weight (g/mol) 538.47
Synonym Diphosphoric Acid P,P,P',P'-Tetrakis(phenylmethyl) Ester, Benzyl Pyrophosphate, Tetrabenzyl Diphosphate
IUPAC Name dibenzyl bis(phenylmethoxy)phosphoryl phosphate
SMILES O=P(OCc1ccccc1)(OCc2ccccc2)OP(=O)(OCc3ccccc3)OCc4ccccc4
Specifications
Quantity 1 g
InChI Formula InChI=1S/C28H28O7P2/c29-36(31-21-25-13-5-1-6-14-25,32-22-26-15-7-2-8-16-26)35-37(30,33-23-27-17-9-3-10-18-27)34-24-28-19-11-4-12-20-28/h1-20H,21-24H2
Formula Weight 538.131
Chemical Name or Material Tetrabenzyl Pyrophosphate