Learn More
Tetra-tert-butyl Orthotitanate, Sigma-Aldrich™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This catalyst offers superior selectivity in transesterification reactions with high regioselectivity. Its sterically hindered structure prevents side reactions, making it valuable for pharmaceutical synthesis under mild conditions.
Specifications
Specifications
| CAS | 3087-39-6 |
| Density | 0.88 g/cm3 at 25°C |
| Boiling Point | 160°C to 165°C/20 hPa |
| Molecular Formula | Ti(OC(CH3)3)4 |
| MDL Number | MFCD00040554 |
| Quantity | 25 mL |
| Synonym | Tetra-tert-butyl orthotitanate, tert-Butyl titanate(IV), Orthotitanic acid tetra-tert-butyl ester, Titanium tetra-tert-butylate, Titanic acid tetra-tert-butyl ester, Tetra-tert-butyl orthotitanate |
| Molecular Weight (g/mol) | 340.32 |
| Grade | Research |