missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TET2 Polyclonal antibody specifically detects TET2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | TET2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of TET2 (NP_001120680.1).,, Sequence:, MEQDRTNHVEGNRLSPFLIPSPPICQTEPLATKLQNGSPLPERAHPEVNGDTKWHSFKSYYGIPCMKGSQNSRVSPDFTQESRGYSKCLQNGGIKRTVSEPSLSGLLQIKKLKQDQKANGERRNFGVSQERNPGESSQPNVSDLSDKKESVSSVAQENAVKDFTSFSTHNCSGPENPELQILNEQEGKSANYHDKNIVLLKNKAVLMPNGATVSASSVEHTHGELLEKTLSQYYPDCVSIAVQKTTSHINAINSQATNELSCEITHPSHTSGQINSAQTSNSELPPKPAAVVSEACDADDADNASKLAAMLNTCSFQKPEQLQQQKSVFEICPSPAENNIQGTTKLASGE |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?