Learn More
Testosterone Enantate EP Impurity B, Mikromol™
Product Code. 17993436
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993436 100 mg 100mg
17934226 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17993436 Supplier Mikromol Supplier No. MM0863.08
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Testosterone Enantate EP Impurity B
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material Testosterone caproate
CAS 10312-45-5
Analyte or Component Names Testosterone caproate
Shipping Condition Room Temperature
Molecular Formula C25 H38 O3
Certifications/Compliance ISO 9001
Synonym Testosterone Caproate, Testosterone Enantate Imp. B (EP), Testosterone Isocaproate Imp. E (EP), 3-Oxoandrost-4-en-17beta-yl Hexanoate, Androst-4-en-3-one, 17-[(1-oxohexyl)oxy]-, (17beta)-, Testosterone, hexanoate (6CI,7CI,8CI), (17beta)-17-[(1-Oxohexyl)oxy]androst-4-en-3-one, 17beta-Hydroxyandrost-4-en-3-one hexanoate, Testosterone 17-caproate, Testosterone caproate, Testosterone Enanthate Related Compound B, Testosterone Enantate Impurity B, Testosterone Isocaproate Impurity E
InChI Formula InChI=1S/C25H38O3/c1-4-5-6-7-23(27)28-22-11-10-20-19-9-8-17-16-18(26)12-14-24(17,2)21(19)13-15-25(20,22)3/h16,19-22H,4-15H2,1-3H3/t19-,20-,21-,22-,24-,25-/m0/s1
SMILES CCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] hexanoate
Molecular Weight (g/mol) 386.57
Quantity 100 mg
Formula Weight 386.2821 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.