missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Antibody (BGN/3G8A4), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18734780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
18734780 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 18734780

Brand: Novus Biologicals NB10062943JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Testosterone
Applications ELISA
Classification Monoclonal
Clone BGN/3G8A4
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone
Host Species Mouse
Immunogen BSA conjugated testosterone-3,-CMO
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Developmental Biology
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.