Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall
Product Code. 17991142 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
3 mL
5 mL
7 mL
10 mL
12 mL
18 mL
22 mL
23 mL
29 mL
35 mL
55 mL
Length (Metric):
75 mm
100 mm
125 mm
150 mm
160 mm
180 mm
Diameter (Metric):
10 mm
12 mm
14 mm
16 mm
18 mm
24 mm
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Diameter (Metric) Unit Size
17991142 7 mL 100 mm 12 mm Pack of 1000
15802265 3 mL 75 mm 10 mm Pack of 100
17901132 5 mL 75 mm 12 mm Pack of 1000
17911122 10 mL 100 mm 14 mm Pack of 1000
17921122 12 mL 100 mm 16 mm Pack of 1000
17971142 18 mL 125 mm 16 mm Pack of 1000
17951142 22 mL 150 mm 16 mm Pack of 1000
17901122 23 mL 160 mm 16 mm Pack of 1000
15802275 29 mL 150 mm 18 mm Pack of 100
17991132 35 mL 180 mm 18 mm Pack of 100
15812275 55 mL 150 mm 24 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 17991142
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from soda lime glass
  • Thick-walled and therefore mechanically very resistant
  • Available with beaded rim or straight rim
  • With round bottom
Specifications
Capacity (Metric) 7 mL
Length (Metric) 100 mm
Material Soda Lime Glass
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No
Diameter (Metric) 12 mm
Product Type Test Tube
Wall Thickness 0.9 mm