Learn More
Hirschmann™ Test Tubes with Round Bottoms
Product Code. 10663582
Change view
Click to view available options
Diameter (Metric):
10 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10663582 10 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10663582 Supplier Hirschmann™ Supplier No. 9270105
Only null left
Add to basket
Add to basket
Manufactured from soda-lime glass.
  • Smooth edge
  • Non-porous surface
  • 75 x 10 mm
Specifications
Description 75 x 10 mm, 0.8 mm thickness
Material Glass
Diameter (Metric) 10 mm