Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint with Graduation
Product Code. 10018289
Change view
Click to view available options
Capacity (Metric):
20 mL
25 mL
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10018289 25 mL Pack of 10
10765262 20 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10018289 Supplier Lenz Laborglasintrumente Supplier No. 05706003
Estimated Shipment: 02-11-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With round bottom
  • With NS socket
  • With PE stopper
  • With scale in high-contrast enamel
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)