Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint, with Graduation
Product Code. 11973571
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Pack of 10
Product Code. Capacity (Metric) unitSize
11973571 30 mL Pack of 10
1 options
Product Code. 11973571
Brand: Lenz Laborglasintrumente 05706004
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With round bottom
  • With NS socket
  • With PE stopper
  • With scale in high-contrast enamel
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 30 mL
Material Borosilicate Glass (3.3)