Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint, with Conical Bottom
Product Code. 10677780
Click to view available options
Diameter (Metric):
17 mm
Length (Metric):
115 mm
120 mm
Closure Type:
Hexagonal
Not Included
Stopper
Unit Size:
Pack of 10
Product Code. 10677780
Brand: Lenz Laborglasintrumente 05705216
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With conical bottom
  • With socket NS 14/23
  • Available without stopper
  • With PE stopper or with hexagonal hollow glass stopper
  • Made of DURAN™ tubing
Specifications
Closure Material Glass
Length (Metric) 120 mm
Material Borosilicate Glass (3.3)
Diameter (Metric) 17 mm
Closure Type Hexagonal
Bottom Shape Conical