Learn More
Velp Scientifica™ Test Tubes for Digestion Units
Product Code. 10777925 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
DK6/48 and UDK series
Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
UDK Series distillation units
Diameter (Metric) Outer:
26 mm
42 mm
48 mm
80 mm
Unit Size:
Each
Pack of 3
Pack of 6
Product Code. For Use With (Equipment) Diameter (Metric) Outer unitSize
10777925 UDK Series distillation units 80 mm Each
11725654 Digesters DK 20/26, DKL 42/26 and UDK Distillation Units 26 mm Pack of 6
11900114 DK6/48 and UDK series 48 mm Each
11715654 - 42 mm Pack of 3
4 options
Product Code. 10777925
Brand: Velp Scientifica™ A00001083
Only null left
Add to basket
Add to basket
Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • Available in different counts
  • Available in different sizes and diameters
Specifications
Product Type Test Tubes
Diameter (Metric) Outer 80 mm
For Use With (Equipment) UDK Series distillation units