Learn More
Grant Instruments™ Test Tube Tray for Grant Shaking Water Baths
Product Code. p-8013373
Change view
Click to view available options
Type:
Flask Tray
Test Tube Tray
Test Tube/Microtube Tray
For Use With (Equipment):
LSB12
LSB18
OLS26
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
15305822 Test Tube Tray LSB18 Each
15395822 Flask Tray OLS26 Each
15385812 Test Tube Tray LSB12 Each
15325822 Test Tube/Microtube Tray OLS26 Each
4 options
Product Code. 15305822
Brand: Grant Instruments™ TS18
Only null left
Add to basket
Add to basket
Holds 3 to 5 SR Racks. For use with Grant Shaking Water Baths
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruemnts™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
Test Tube Tray for Grant Shaking Water Baths
  • TS12: Holds up to 3 SR Racks for LSB12 Model Aqua Pro Shaking Water Bath
  • TS18: Holds up to 5 SR Racks for LSB18 Model Aqua Pro Shaking Water Bath
  • TS26: Holds up to 5 SR Racks for OLS26 Model Aqua Pro Shaking Water Bath
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LSB18
Type Test Tube Tray
Holds Up to 3 SR Racks
Material Stainless Steel