Learn More
Lenz Laborglasintrumente Test Tube Sprayers
Product Code. 11993531
Change view
Click to view available options
Description:
Test tube and sprayer head
Test tube for head
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11993531 Test tube for head Each
11586904 Test tube and sprayer head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11993531 Supplier Lenz Laborglasintrumente Supplier No. 05470606
  • ONLINE EXCLUSIVE PRICE
    103.00 NOK
    86.28 NOK / Pack of 1
    Save 16.72 NOK
    16% Off
Estimated Shipment: 14-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Complete sprayer consisting of: Test tube and sprayer head (made of DURAN™ tubing)
  • With clamp and rubber bulb
  • Packaging unit: 1 piece
Specifications
Capacity (Metric) 6 mL
Description Test tube for head
Standard Taper Joint NS 14/23