Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Product Code. 11875820 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Holds:
Tubes (1.5 mL)
Tubes (10 to 13 mm)
Tubes (14 to 16 mm), Centrifuge Tubes (15 mL)
Tubes (17 to 20 mm)
Tubes (21 to 25 mm)
Tubes (26 to 30 mm)
Tubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Array:
3 x 3
3 x 8 array
4 x 10
4 x 4
4 x 5
4 x 6
6 x 12
6 x 6
8 x 12
Unit Size:
Each
Product Code. Color Holds Array unitSize
11875820 Blue Tubes (1.5 mL) 8 x 12 Each
11719518 Blue Tubes (1.5 mL) 4 x 6 Each
11436358 Red Tubes (10 to 13 mm) 6 x 6 Each
10174383 Red Tubes (10 to 13 mm) 6 x 12 Each
10794214 Orange Tubes (14 to 16 mm), Centrifuge Tubes (15 mL) 6 x 6 Each
10031913 Orange Tubes (14 to 16 mm), Centrifuge Tubes (15 mL) 6 x 12 Each
11476358 White Tubes (17 to 20 mm) 4 x 5 Each
10417883 White Tubes (17 to 20 mm) 4 x 10 Each
11486358 Blue Tubes (21 to 25 mm) 4 x 4 Each
11426358 Blue Tubes (21 to 25 mm) 4 x 10 Each
11466358 Green Tubes (26 to 30 mm) 3 x 8 array Each
11496358 Green Tubes (26 to 30 mm), Centrifuge Tubes (50 mL) 3 x 3 Each
12 options
Product Code. 11875820
Brand: Thermo Scientific™ 30190
Only null left
Add to basket
Add to basket
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
TRUSTED_SUSTAINABILITY
Specifications
Description Blue; 1.5 mL; 8 x 12 array
Color Blue
Holds Tubes (1.5 mL)
Positions 96
For Use With (Equipment) MaxQ™ Shaker Platform
Product Type Test Tube Rack
Array 8 x 12