Learn More
Fisherbrand™ Test Tube Rack for Fisherbrand™ Heated Circulating Baths
Product Code. 17581916 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
102 x 0.5 mL Tubes
14 x 30 mm Tubes
23 x 24 mm Tubes
36 x 16 to 19 mm Tubes
65 x 10 to 13 mm Tubes
75 x 1.5 mL Tubes
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
17581916 14 x 30 mm Tubes Each
17571916 23 x 24 mm Tubes Each
17561916 36 x 16 to 19 mm Tubes Each
17551916 65 x 10 to 13 mm Tubes Each
17591916 75 x 1.5 mL Tubes Each
17501926 102 x 0.5 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17581916
Only null left
Add to basket
Add to basket
These test tube racks accommodate a variety of tube sizes and capacities.
  • For use with a number of Fisherbrand™ Heating Circulators and Heated Circulating Baths
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Test Tubes
Holds 14 x 30 mm Tubes