Learn More
Julabo™ Test Tube Rack
Product Code. 15430632
Click to view available options
No. of Positions:
21
30
42
60
90
Unit Size:
Each
Product Code. 15430632
Brand: Julabo™ 9970300
Only null left
Add to basket
Add to basket
Test tube rack made of high grade plastic or stainless steel
Specifications
Description Test tube racks for water baths
Temperature Range Up to +100°C
Material High grade plastic
No. of Positions 60
Size 100 x 16/17 mm dia.