Learn More
OHAUS™ Test Tube Rack
Product Code. 15792529 Shop All Ohaus Products
Change view
Click to view available options
Positions:
12
17
21
24
33
35
40
48
60
63
90
Holds:
Tubes (10 to 13 mm)
Tubes (10 to 14 mm)
Tubes (14 to 16 mm)
Tubes (15 mL)
Tubes (16 mm)
Tubes (16 to 20 mm)
Tubes (18 to 20 mm)
Tubes (20 mm)
Tubes (21 to 25 mm)
Tubes (22 to 25 mm)
Tubes (25 mm)
Tubes (30 mm)
Tubes (5 mL)
Unit Size:
Each
Product Code. Positions Holds unitSize
15792529 35 Tubes (18 to 20 mm) Each
15722539 12 Tubes (5 mL) Each
15752529 17 Tubes (5 mL) Each
15742529 21 Tubes (21 to 25 mm) Each
15712529 21 Tubes (30 mm) Each
15702539 24 Tubes (22 to 25 mm) Each
15702529 24 Tubes (25 mm) Each
15732529 33 Tubes (16 to 20 mm) Each
15712539 35 Tubes (15 mL) Each
15792519 40 Tubes (20 mm) Each
15722529 48 Tubes (10 to 14 mm) Each
15782529 48 Tubes (14 to 16 mm) Each
15782519 60 Tubes (16 mm) Each
15772529 63 Tubes (10 to 13 mm) Each
15772519 90 Tubes (30 mm) Each
15 options
Product Code. 15792529
Brand: OHAUS™ 30400117
Only null left
Add to basket
Add to basket
OHAUS™ Test Tube Rack designed to securely hold tubes in place.
  • Constructed of stainless steel
  • For use with tubes of various sizes
TRUSTED_SUSTAINABILITY
Specifications
Description Test Tube Rack 18-20 mm Diameter
Autoclavable Autoclavable
Color Blue
Holds Tubes (18 to 20 mm)
Material PVC-coated Steel
Positions 35
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Size 18 to 20 mm
Dimensions (L x W x H) 12.7 x 17.8 x 10.2 cm
Length (Metric) 12.70 cm
Length (English) 5.00 in.
Width (English) 7.00 in.
Height (English) 4.00 in.
Depth (English) 5.00 in.
Depth (Metric) 12.70 cm
Diameter (Metric) Opening 20 mm
Product Type Test Tube Rack
Shelves 1
Height (Metric) 10.200 cm
Array 5 x 7
Width (Metric) 17.80 cm