Description
- Constructed of stainless steel
- For use with tubes of various sizes
Specifications
Specifications
| Description | Test Tube Rack 21-25 mm Stationary |
| Autoclavable | Autoclavable |
| Color | Blue |
| Holds | Tubes (21 to 25 mm) |
| Material | PVC-coated Steel |
| Positions | 21 |
| Shape | Rectangular |
| Stackable | Yes, When Empty |
| Sterility | Non-sterile |
| Model | Stationary |
| Show More |