Learn More
OHAUS™ Test Tube Rack
Product Code. 15702529 Shop All Ohaus Products
Click to view available options
Positions:
12
17
21
24
33
35
40
48
60
63
90
Holds:
Tubes (10 to 13 mm)
Tubes (10 to 14 mm)
Tubes (14 to 16 mm)
Tubes (15 mL)
Tubes (16 mm)
Tubes (16 to 20 mm)
Tubes (18 to 20 mm)
Tubes (20 mm)
Tubes (21 to 25 mm)
Tubes (22 to 25 mm)
Tubes (25 mm)
Tubes (30 mm)
Tubes (5 mL)
Unit Size:
Each
Product Code. 15702529
Brand: OHAUS™ 30400108
Only null left
Add to basket
Add to basket
OHAUS™ Test Tube Rack designed to securely hold tubes in place.
  • Constructed of stainless steel
  • For use with tubes of various sizes
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pivoting Test Tube Rack
Description Test Tube Rack 25 mm Pivoting
Array 3 x 8
Autoclavable Autoclavable
Color Blue
Holds Tubes (25 mm)
Material Stainless Steel
Positions 24
Shape Rectangular
Shelves 2 Tiers
Stackable No
Sterility Non-sterile
Model Pivoting
Size 25 mm
Dimensions (L x W x H) 12.7 x 27.7 x 12.7 cm
Length (Metric) 12.70 cm
Length (English) 5.00 in.
Width (Metric) 27.70 cm
Width (English) 10.90 in.
Height (Metric) 12.700 cm
Height (English) 5.00 in.
Depth (English) 5.00 in.
Depth (Metric) 12.70 cm
Diameter (Metric) Opening 25 mm