Learn More
Memmert™ Test Tube Rack
Test Tube Rack designed for use with a Memmert™ Water Bath.
Brand: Memmert™ WTB50B50657
Additional Details : Weight : 2.00000kg
Description
- Water Bath Accessory
Specifications
WTB50 Water Baths | |
66 Test Tubes (2 Rows) | |
12 mm |
Test Tube Rack | |
Stainless Steel |