Learn More
Labconco™ Test Tube/Microcentrifuge Tube Rack: FreeZone™ Stoppering or Bulk Tray Dryer Accessory
Click to view available options
Capacity (Metric):
70 Tubes
Unit Size:
Each
Description
Rectangular Test Tube/Microcentrifuge Rack
- Holds up to 70 12-13 mm tubes
- Made of type 304 stainless steel
- Includes bottom and adapter plate for (70) microcentrifuge tubes
- Ideal for use with FreeZone™ Tray Dryer, Bulk Tray Dryer, Stoppering Tray Dryer
- Dimensions: 7.4 in. w x 11.5 in. d x 2.5 in. h
Specifications
Specifications
| Type | Tube Rack |
| Capacity (Metric) | 70 Tubes |
| No. of Shelves | 0 |
| Material | Stainless Steel (304) |
| For Use With (Equipment) | FreeZone™ Tray Dryer, Bulk Tray Dryer, or Stoppering Tray Dryer |
| Includes | Stainless steel rack and 1 stainless steel microcentrifuge tube adapter plate |
| Width (English) | 7.4 in. |
| Height (English) | 2.5 in. |
| Length (English) | 11.5 in. |
| Shipping Weight (English) | 3.5 lb. |
| Show More |