Learn More
Biochrom Test Tube Holder for Ultrospec 7000/8000/9000
Product Code. 11633024 Shop All Harvard Bioscience Products
Change view
Click to view available options
Type:
Test Tube Holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11633024 Test Tube Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11633024 Supplier Biochrom Supplier No. 80710011
  • ONLINE EXCLUSIVE PRICE
    11515.00 NOK
    10576.79 NOK / Pack of 1
    Save 938.21 NOK
    8% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Test Tube Holder designed to hold test tubes 10 to 24 mm in diameter and up to 160 mm tall in the Ultrospec 7000, 8000 and 9000 spectrophotometers. Replaces the standard single cell holder. Easily installed by the operator with only one thumb screw to remove at the front of the cell compartment.
  • For Ultrospec 7000/8000/9000
  • Fits 10 to 24 mm tube diameters up to 160 mm tube height
  • Fits to standard sample compartment baseplate
  • Can be changed in seconds
Specifications
For Use With (Equipment) Ultrospec 7000/8000/9000
Type Test Tube Holder
Diameter (English) 0.4 to 0.9 in.
Diameter (Metric) 10 to 24 mm
For Use With (Application) Holds 10 to 24 mm dia. x 160 mm H Test Tubes
Material Carbon Fiber