Learn More
OHAUS™ Test Tube Holder
Product Code. 15742639 Shop All Ohaus Products
Change view
Click to view available options
Holds:
13 x 22 to 25 mm Test Tubes
18 x 19 to 21 mm Test Tubes
20 x 15 to 18 mm Test Tubes
34 x 12 to 13 mm Test Tubes
4 x 26 to 29 mm Test Tubes
52 x 0.5 mL Microtubes
Unit Size:
Each
Product Code. Holds unitSize
15742639 18 x 19 to 21 mm Test Tubes Each
15732639 20 x 15 to 18 mm Test Tubes Each
15782629 52 x 0.5 mL Microtubes Each
15752639 13 x 22 to 25 mm Test Tubes Each
15762639 4 x 26 to 29 mm Test Tubes Each
15722639 34 x 12 to 13 mm Test Tubes Each
6 options
Product Code. 15742639
Brand: OHAUS™ 30400222
Only null left
Add to basket
Add to basket
Hands free mixing designed to fit on the Heavy Duty Vortex Mixer. Easily attached to the top of mixer and is magnetically secured.
  • Use with Heavy Duty Vortex Mixer
TRUSTED_SUSTAINABILITY
Specifications
Holds 18 x 19 to 21 mm Test Tubes
Tube Size 19 to 21 mm
Product Type Foam Insert
Material Foam
Description Foam Insert For (18) 19-21 mm Tubes
For Use With (Equipment) Vortex Mixer
Packaging Quantity 1 Ea.