Learn More
OHAUS™ Test Tube Holder
Shop All Ohaus Products
Click to view available options
Holds:
13 x 22 to 25 mm Test Tubes
18 x 19 to 21 mm Test Tubes
20 x 15 to 18 mm Test Tubes
34 x 12 to 13 mm Test Tubes
4 x 26 to 29 mm Test Tubes
52 x 0.5 mL Microtubes
Unit Size:
Each
Description
- Use with Heavy Duty Vortex Mixer
Specifications
Specifications
| Holds | 34 x 12 to 13 mm Test Tubes |
| Tube Size | 12 to 13 mm |
| Product Type | Foam Insert |
| Material | Foam |
| Description | Foam Insert For (34) 12-13 mm Tubes |
| For Use With (Equipment) | Vortex Mixer |
| Packaging Quantity | 1 Ea. |