Learn More
Reitenspiess™ Test Tube Brush
Product Code. 11361265
Change view
Click to view available options
Length (Metric):
100 mm
190 mm
230 mm
Diameter (Metric) Bristle:
16 mm
26 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Bristle Unit Size
11361265 190 mm 16 mm Each
11371265 - - Each
11351265 230 mm 26 mm Each
11327494 - - Each
11337494 100 mm - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11361265 Supplier Reitenspiess™ Supplier No. 12210301
  • ONLINE EXCLUSIVE PRICE
    214.80 NOK
    30.04 NOK / Pack of 1
    Save 184.76 NOK
    86% Off
In Stock
Add to basket
Edge
Add to basket
Use a Reitenspiess™ Test Tube Brush for cleaning, disinfecting and sterilizing test tubes as necessary in the laboratory. .
  • Natural bristle
  • Various lengths available
  • Various diameters
Specifications
Product Type Test Tube Brush
Diameter (Metric) Bristle 16 mm
Length (Metric) 190 mm
Quantity 1 Ea.